missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PVRIG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21448-1-AP-150UL
This item is not returnable.
View return policy
Description
Cell surface receptor for NECTIN2. May act as a coinhibitory receptor that suppresses T-cell receptor-mediated signals. Following interaction with NECTIN2, inhibits T-cell proliferation. Competes with CD226 for NECTIN2-binding.Specifications
PVRIG | |
Polyclonal | |
Unconjugated | |
PVRIG | |
C7orf15, CD112R, PVRIG, Transmembrane protein PVRIG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79037 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6DKI7 | |
PVRIG | |
PVRIG Fusion Protein Ag13815 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PVRIG Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction