missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ 40mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 40mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
2300.00 SEK - 2515.00 SEK
Specifications
Capacity (Metric) | 50 mL |
---|---|
Diameter (Metric) Stem | 8mm |
Material | Borosilicate Glass, Borosilicate Glass |
Height (Metric) | 170 mm |
Diameter (Metric) Disc | 40mm |
Product Code | Brand | Pore Size | Porosity | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Porosity | Includes | Price | Quantity & Availability | |||
12673212
|
Pyrex
3630/02 |
100 to 160μm | 1 | Integrated filter plate sintered Glass disc |
2390.00 SEK
Pack of 5 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
12683212
|
Pyrex
3630/04 |
40 to 100μm | 2 | Integrated filter plate sintered Glass disc |
2390.00 SEK
Pack of 5 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
11991548
|
Pyrex
3630/06 |
10 to 16μm | 3 | Integrated filter plate sintered Glass disc |
2395.00 SEK
Pack of 5 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||
12603222
|
Pyrex
3630/08 |
10 to 16μm | 4 | Integrated filter plate sintered Glass disc |
2365.00 SEK
Pack of 5 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
12692132
|
Pyrex
3630/01 |
160 to 250μm | 0 | Integral sintered Glass discs |
2300.00 SEK
Pack of 5 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
12602142
|
Pyrex
3630/10 |
1.0 to 1.6μm | 5 | Integrated filter plate sintered Glass disc |
2515.00 SEK
Pack of 5 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
Description
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
- Choice of disc diameters and porosities
Specifications
50 mL | |
Borosilicate Glass, Borosilicate Glass | |
40mm | |
Buchner Funnel |
8mm | |
170 mm | |
100mm |