Learn More
PYREX™ 95mm Diameter Glass Buchner Filter Funnel
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 95mm Glass Buchner Filter Funnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
Brand: Pyrex 3710/02
24985.00 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.42000kg
Description
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 95mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
- Choice of disc diameters and porosities
Specifications
500 mL | |
Borosilicate Glass, Borosilicate Glass | |
For easy filtering solutions | |
Integrated filter plate sintered Glass disc | |
1 | |
Buchner Funnel |
19mm | |
250 mm | |
95mm | |
115mm | |
100 to 160μm |