Learn More
Pyrex™ Aspirator Bottle with Ground Glass Side Socket
Manufactured from borosilicate glass
Brand:  Pyrex 1541/16D
1904.75 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 2.27000kg
 View more versions of this product
Product Code. 11347884
  • 2005.00 SEK / Each
Estimated Shipment: 21-10-2024
to see stock.
Add to basket
Description
Description
Manufactured from borosilicate glass
  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Unground top neck to accept rubber stoppers
Specifications
Specifications
5000 mL
Borosilicate Glass
Aspirator Bottle
44 mm
318 mm
181 mm