missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Narrow-Mouth Reagent Bottle
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
15,000 mL
20,000 mL
2000 mL
250 mL
500 mL
5000 mL
Description
Manufactured from Pyrex™ superior strength borosilicate glass. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles are ideal for storing chemicals in the laboratory. Use rubber stoppers (not included) to secure.
- Unground neck for use with rubber stoppers (not supplied)
- Borosilicate glass provides strong chemical resistance
- Complies with ISO 4796

Specifications
Specifications
Capacity (Metric) | 250 mL |
Material | Borosilicate Glass |
Quantity | 10pack |
Certified | Yes |
Neck Finish | Unground |
Certifications/Compliance | ISO 4796 |
Closure Included | No |
Mouth | Narrow |
Height (Metric) | 131 mm |
Product Type | Reagent Bottle |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrex™ Narrow-Mouth Reagent Bottle >
Spot an opportunity for improvement?Share a Content Correction