Learn More
Pyrex™ Wide Neck Reagent Bottle
3557.75 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Use promo code "23259" to get your promotional price.
Click to view available options
Capacity (Metric):
10 L
Description
- Complies with ISO 4796
- Borosilicate glass with unground neck
- For use with rubber stoppers (not supplied)
Specifications
For Use With (Equipment) | Rubber Stoppers |
Product Type | Reagent Bottle |
Capacity (Metric) | 10 L |
Material | Borosilicate Glass |
Certifications/Compliance | ISO 4796 |
Mouth | Unground Wide |