missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal 5-Phosphate (Monohydrate), Fisher BioReagents

640.00 SEK - 4920.00 SEK

Chemical Identifiers

CAS 54-47-7
Molecular Formula C8H10NO6P
Molecular Weight (g/mol) 247.14
MDL Number MFCD00006333
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
Synonym pyridoxal phosphate, codecarboxylase, pyridoxal 5-phosphate, pyridoxal 5'-phosphate, pyridoxal p, biosechs, hairoxal, pyromijin, vitazechs, hiadelon
PubChem CID 1051
ChEBI CHEBI:18405
IUPAC Name (4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10516204
View Documents
Fisher BioReagents
BP2676-1
1 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10516394
View Documents
Fisher BioReagents
BP2676-5
5 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10033284
View Documents
Fisher BioReagents
BP2676-25
25 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • PLP is useful for modifying lysyl and valyl residues and functions as a coenzyme in transamination reactions by forming a Schiff-base linkage with lysine groups on aminotransferase
  • PLP is an active form of vitamin B6
  • Also serves as a coenzyme in some decarboxylation and deamination reactions
Specifications

Chemical Identifiers

54-47-7
247.14
NGVDGCNFYWLIFO-UHFFFAOYSA-N
1051
(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate
C8H10NO6P
MFCD00006333
pyridoxal phosphate, codecarboxylase, pyridoxal 5-phosphate, pyridoxal 5'-phosphate, pyridoxal p, biosechs, hairoxal, pyromijin, vitazechs, hiadelon
CHEBI:18405
CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Specifications

54-47-7
White
≥98%
MFCD00006333
pyridoxal phosphate, codecarboxylase, pyridoxal 5-phosphate, pyridoxal 5'-phosphate, pyridoxal p, biosechs, hairoxal, pyromijin, vitazechs, hiadelon
CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O
247.14
CHEBI:18405
≥98%
Pyridoxal 5-Phosphate, Monohydrate
140°C
Conforms to reference
C8H10NO6P
15, 8092
NGVDGCNFYWLIFO-UHFFFAOYSA-N
(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate
1051
247.14
Powder or Solid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store below 0°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.