Learn More
Tocris Bioscience™ Pyrintegrin
Enhances survival of human ESCs following enzymatic dissociation
Brand:  Tocris Bioscience™ 4978/10
Product Code. 18709258
  • 4920.00 SEK / 10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Pyrintegrin enhances survival of human embryonic stem cells (hESCs) following enzymatic dissociation. Thought to enhance cell-ECM adhesion and activate integrin signaling. Pyrintegrin increases adhesion of hESCs to matrigel- and laminin-coated plates, but not to gelatin-coated plates. Also podocyte-protective agent and prevents LPS-induced proteinuria in mice.
Specifications
Specifications
>98%
Pyrintegrin
Pyrintegrin
10 mg
451.54