Learn More
Pyromellitic acid, 96%, Thermo Scientific Chemicals
440.22 SEK - 1455.55 SEK
Chemical Identifiers
CAS | 89-05-4 |
---|---|
Molecular Formula | C10H6O8 |
Molecular Weight (g/mol) | 254.15 |
MDL Number | MFCD00002471 |
InChI Key | CYIDZMCFTVVTJO-UHFFFAOYSA-N |
Synonym | pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene |
PubChem CID | 6961 |
ChEBI | CHEBI:45165 |
IUPAC Name | benzene-1,2,4,5-tetracarboxylic acid |
SMILES | C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11496296
|
Thermo Scientific Alfa Aesar
B23099.22 |
100 g |
440.22 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11406306
|
Thermo Scientific Alfa Aesar
B23099.36 |
500 g |
1455.55 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pyromellitic acid is an eluent used in anion chromatography and forms complexes with magnesium and calcium ions. It is used as a curing agent, as an intermediate for powder coating and a cross-linking agent for alkyd resin. It is involved in the synthesis of polyimide and pyromellitic octyl.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyromellitic acid is an eluent used in anion chromatography and forms complexes with magnesium and calcium ions. It is used as a curing agent, as an intermediate for powder coating and a cross-linking agent for alkyd resin. It is involved in the synthesis of polyimide and pyromellitic octyl.
Solubility
Soluble in alcohol.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
89-05-4 | |
254.15 | |
CYIDZMCFTVVTJO-UHFFFAOYSA-N | |
6961 | |
benzene-1,2,4,5-tetracarboxylic acid |
C10H6O8 | |
MFCD00002471 | |
pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene | |
CHEBI:45165 | |
C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O |
Specifications
89-05-4 | |
C10H6O8 | |
14,8004 | |
Soluble in alcohol. | |
C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O | |
254.15 | |
CHEBI:45165 | |
96% |
281°C to 285°C | |
MFCD00002471 | |
pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene | |
CYIDZMCFTVVTJO-UHFFFAOYSA-N | |
benzene-1,2,4,5-tetracarboxylic acid | |
6961 | |
254.15 | |
Pyromellitic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 201-879-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only