missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ QO 58
Kv7 channel opener
2700.79 SEK - 10709.22 SEK
Specifications
Chemical Name or Material | 5-(2,6-Dichloro-5-fluoro-3-pyridinyl)-3-phenyl-2-(trifluoromethyl)-pyrazolo[1,5-a]pyrimidin-7(4H)-one |
---|---|
CAS | 1259536-62-3 |
Target | Voltage-gated Potassium Channel Activators |
Molecular Formula | C18H8Cl2F4N4O |
Formula Weight | 443.18 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702158
|
Tocris Bioscience™
5083/10 |
10 mg |
2700.79 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18710939
|
Tocris Bioscience™
5083/50 |
50 mg |
10709.22 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kv7 channel opener (EC50 values are 0.6, 1.0, 5.2 and 7.0 μM for Kv7.4, Kv7.2, Kv7.3/7.5 and Kv7.1, respectively). Increases the pain threshold of neuropathic pain in a sciatic nerve CCI in vivo model.Specifications
5-(2,6-Dichloro-5-fluoro-3-pyridinyl)-3-phenyl-2-(trifluoromethyl)-pyrazolo[1,5-a]pyrimidin-7(4H)-one | |
Voltage-gated Potassium Channel Activators | |
443.18 |
1259536-62-3 | |
C18H8Cl2F4N4O | |
>98% |
Safety and Handling
Recommended Storage : Store at 4°C