Learn More
Socorex™ Qualitix™ Micropipette Tips, 10μL
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 10μL also come in a variety of shapes to facilitate fluid transfer.
Brand: Socorex™ 3090.0010PR
This item is not returnable.
View return policy
Description
- Silicone-free resin construction.
- Maximal liquid recovery.
- Available in sterile or non-sterile racks.
- Racks and tips are autoclavable.
- Sterile tips may be autoclaved if required.
- Protease free certified.
Specifications
10 μL | |
Non-filtered | |
Autoclavable | |
96/Rack | |
Single-use | |
Qualitix |
10μL | |
Non-sterile | |
Ultra-micro, Fine Point, Low Binding | |
12/PK, 10PK/CS | |
Universal Fit Micro |