Learn More
Socorex™ Qualitix™ Micropipette Tips, 10μL
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 10μL also come in a variety of shapes to facilitate fluid transfer.
Brand:  Socorex™ 3090.0010PR
 View more versions of this product
Product Code. 15720669
  • 1165.00 SEK / Pack of 1152
Estimated Shipment: 03-10-2024
to see stock.
Explore more special offers
Description
  • Silicone-free resin construction.
  • Maximal liquid recovery.
  • Available in sterile or non-sterile racks.
  • Racks and tips are autoclavable.
  • Sterile tips may be autoclaved if required.
  • Protease free certified.
Specifications
Specifications
10 μL
Non-filtered
Autoclavable
96/Rack
Single-use
Qualitix
10μL
Non-sterile
Ultra-micro, Fine Point, Low Binding
12/PK, 10PK/CS
Universal Fit Micro