Learn More
Munktell Filtrak™ Quantitative Ashless Papers
Recommended for use where a high purity is required for quantitative analysis. Munktell Filtrak™ Quantitative Ashless Papers; Circle are acid-washed to remove impurities and are made using pure cellulose with an alpha-cellulose content of almost 100%.
Brand:  Munktell Filtrak™ 100105
19.63 SEK valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Additional Details : Weight : 0.02700kg
 View more versions of this product
Product Code. 11322974
  • 140.00 SEK / Pack of 100
In Stock
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.
Description
Description
  • Very fast filtering, open fiber matrix
  • Fast filtering, open to semi-open fiber matrix
  • Medium to fast filtering, semi-open to open fiber matrix
  • Medium to slow filtering, medium-tight fiber matrix
  • Very slow filtration, very tight fiber matrix
Specifications
Specifications
Fine crystalline
5 to 6 μm
OOK
Cellulose
20s/10mL
55 mm