missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 12K Flex OpenArray™ Calibration Kit

Offered in order to maintain the QuantStudio™ 12K Flex Real Time PCR System OpenArray™ block type.

Brand:  Applied Biosystems™ 4478601

Product Code. 15978840

  • 12170.00 SEK / 1 unit

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The calibration kit contains FAM™ dye, as well as a calibration starter kit. The calibration starter kit contains the calibration plaque for background and uniformity, as well as four empty cases for dye loading. Calibration is recommended on a yearly basis. Please see your instrument maintenance guide for full instructions on use.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dye Calibration, Background Calibration, Uniformity Calibration
QuantStudio™ 12k Flex
QuantStudio™
1 kit
1 Bottle Fam Dye, store at -20°C
1 Calibration Starter Kit, store at room temperature
FAM
PCR Calibration Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.