Learn More
Hellma™ Quartz Suprasil Ultra-Micro Fluorescence Cells
Brand:  Hellma™ 105-251-85-40
Product Code. 14855081
  • 8395.00 SEK / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Specifications
Ultra-micro Fluorescence Cell
Quartz Suprasil
3 x 3 mm
12.5 mm
45 μL
1 Each
45 mm