man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Specialty Lab Glassware
Flow Cells
Hellma™ Quartz UV Flow Cell
12637201
Print
Hellma™ Quartz UV Flow Cell
Capacity: 120μL
Brand:
Hellma™ 170-000-1-40
View more versions of this product
Product Code.
12637201
8375.00 SEK
/ Each
Quantity
Estimated Shipment: 02-05-2025
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Specifications
Type
UV Flow Cell
Capacity (Metric)
120 μL
Material
Quartz
Path Length
1 mm