Learn More
Systec™ Quick Cooling
Use this cooling system with Systec™ autoclaves. Systec Quick Cooling Water Systems reduce cool-down time by up to 90%.
Brand:  Systec™ 6057
Product Code. 10395253
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Cooling system for sterilizing liquids in bottles
  • Reduces the cool-down time by up to 90% when compared to self-cooling
Specifications
Specifications
Systec autoclaves
Quik Cooling