Learn More
Quickfit™ Funnel with Ground Glass Cone
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Quickfit™ Funnel with Ground Glass Cone is designed for transferring powders into Quickfit™ flasks and vessels.
Marque: Quickfit CF5/5
617.50 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Informations supplémentaires : Weight : 0.21600kg
Description
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Quickfit™ Funnel with Ground Glass Cone is designed for transferring powders into Quickfit™ flasks and vessels.
- Designed for transferring powders into Quickfit™ flasks and vessels
- With Quickfit™ ground glass cone
- Manufactured from chemically resistant Pyrex™ Borosilicate Glass
Spécification
105mm | |
34/35 | |
Powder Funnel |
Borosilicate Glass, Borosilicate Glass | |
Quickfit™ Ground Glass Cone |