missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Glass Shaft Anchor
Ideal for use with Quickfit™ wide-neck reaction flasks FR:F series.
Brand: Quickfit ST1/8
3481.75 SEK valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Designed for low speed operation
- Paddle closely follows the contour of the flask walls
- Useful where the build-up of solid material on the walls is likely
- Minimum neck size required for the stirrer is 100mm
Specifications
11 mm | |
For low speed operation | |
750mm |
Quickfit™ wide-neck reaction flasks FR:F series | |
135mm | |
Stirrer Shaft Anchor |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction