Learn More
Quickfit™ Sealing Disc
107.35 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Use promo code "23259" to get your promotional price.
Click to view available options
Quantity:
5/Pk
Description
- For ML1 microscale kit and microscale dropping funnel
Specifications
Diameter (Metric) | 20 mm |
For Use With (Equipment) | ML1 microscale kit |
Quantity | 5/Pk |