missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Hydroxy-2-phenylpropionic acid, 98+%, Thermo Scientific Chemicals

584.00 SEK - 2445.00 SEK

Chemical Identifiers

CAS 3966-30-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00067699
InChI Key NWCHELUCVWSRRS-SECBINFHSA-N
Synonym r---2-hydroxy-2-phenylpropionic acid, 2r-2-hydroxy-2-phenylpropanoic acid, unii-h38dkr9931, r-2-hydroxy-2-phenylpropanoic acid, phenyllactic acid, atrolactic acid,-, 2-hydroxy-2-phenylpropanoic acid #, r---atrolactic acid, r-phenyl lactic acid, r-alpha-hydroxy-alpha-methylbenzeneacetic acid
PubChem CID 736858
IUPAC Name (2R)-2-hydroxy-2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)(C(=O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11302636
SDS View Documents
Thermo Scientific Alfa Aesar
L08644.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11392626
SDS View Documents
Thermo Scientific Alfa Aesar
L08644.03
1 g
2445.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the asymmetric synthesis. Part III. Stereospecific synthesis of (R)-2-hydroxy-2-phenylpropionic acid, and (R)- and (S)-2-cyclohexyl-2-hydroxy-2-phenylacetic acid. Configurational relationship between (R)(-)-2-hydroxy-2-phenylpropionic acid and (S)(+)-2-phenylpropionic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the asymmetric synthesis. Part III. Stereospecific synthesis of (R)-2-hydroxy-2-phenylpropionic acid, and (R)- and (S)-2-cyclohexyl-2-hydroxy-2-phenylacetic acid. Configurational relationship between (R)(-)-2-hydroxy-2-phenylpropionic acid and (S)(+)-2-phenylpropionic acid.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(-)-2-Hydroxy-2-phenylpropionic acid, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.