missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Methyl-2-butyl isocyanate, 97%, Thermo Scientific™

Product Code. 15449149
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 745783-84-0
Density 0.88
Boiling Point 124°C
Flash Point 28°C (82°F)
Molecular Formula C6H11NO
Refractive Index 1.411
MDL Number MFCD05664086
Quantity 1 g
UN Number UN3080
Sensitivity Moisture sensitive; May form a precipitate
Synonym r---3-methyl-2-butyl isocyanate, 2r-2-isocyanato-3-methylbutane, r---3-methyl-2-butylisocyanate
InChI Key UCSWKSXDJYECKQ-ZCFIWIBFSA-N
Optical Rotation −29° (c=1 in Dichloromethane)
SMILES CC(C)C(C)N=C=O
IUPAC Name (2R)-2-isocyanato-3-methylbutane
Molecular Weight (g/mol) 113.16
PubChem CID 7018273
Formula Weight 113.16
Percent Purity 97%
Chemical Name or Material (R)-(-)-3-Methyl-2-butyl isocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.