Learn More
R&D Systems™ CryoDefend-Cell Lines Media
Media for defined, protein-free cryopreservation of cultured cells
Brand:  R&D Systems™ CCM019
Encompass
Product Code. 15700073
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: 5 x 10mL vials CryoDefend-Stem Cells Media
This item is not returnable. View return policy
Description
Description
Cryopreservation can be used to maintain cells at a low passage number and to reduce the risk of contamination by microbes and other cell types. The viability of the recovered cells depends on cell density, the type of cryoprotectant used, the method of cooling, and the amount of the cryoprotectant that remains in the culture after the cells have been thawed.
Contains a defined serum substitute as well as an optimized concentration of a cryopreservative that increases the recovery and viability of healthy cells compared to conventional freezing media
This product contains
DMSO and is ready for use
Reduces experimental variability by using fully defined media
Suitable for all cell types