Learn More
Tocris Bioscience™ Deltarasin
High affinity PDEδ-KRAS interaction inhibitor
Brand: Tocris Bioscience™ 5424/10
This item is not returnable.
View return policy
Description
Deltarasin is a high affinity PDEδ-KRAS interaction inhibitor (Kd = 41 nM); binds to PDEδ. Reduces proliferation and induces cell death of KRAS-dependent Panc-Tu-1 and Capan-1 pancreatic cancer cells. Attenuates tumor growth of Panc-Tu-1 xenografts in mice.Specifications
Other Ras GTPase Compounds | |
2-[4-[(2S)-2-(2-Phenyl-1H-benzimidazol-1-yl)-2-(4-piperidinyl)ethoxy]phenyl]-1-(phenylmethyl)-1H-benzimidazole trihydrochloride | |
Soluble to 100mM in water and to 100mM in DMSO | |
Bioactive Small Molecule |
1440898-82-7 | |
10 mg | |
Solid |
Research Use Only. Not For Human or Veterinary Use