Learn More
R&D Systems™ Human 5-HT3A (NP_000860) VersaClone cDNA
5-Hydroxytryptamine Receptor 3A
Brand:  R&D Systems™ RDC0435
Product Code. 18441260
  • 3340.00 SEK / 10µg
Explore more special offers
Description
Description
The Human 5-HT3A (NP_000860) VersaClone cDNA clone contains the complete ORF for 5-HT3A, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The 5-HT3A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
h5-HT3A
10 μg at 0.2 μg/μL
1468bp
Human
NP_000860
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Special Offers
For Reasearch Use Only; Not for Use in Humans