Learn More
R&D Systems™ Human TREM2 (NP_061838) VersaClone cDNA
Triggering Receptor Expressed on Myeloid Cells 2
Brand: R&D Systems™ RDC0861
This item is not returnable.
View return policy
Description
The Human TREM-2 (NP_061838) VersaClone cDNA clone contains the complete ORF for TREM-2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TREM-2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hTREM2 | |
10 μg at 0.2 μg/μL | |
706bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
NP_061838 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2 triggering receptor expressed on myeloid cells 2 A, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
Human |
For Reasearch Use Only; Not for Use in Humans