Learn More
R&D Systems™ Rat OX40/TNFRSF4 (NP_037181) VersaClone cDNA
Tumor Necrosis Factor Receptor Superfamily, Member 4
Brand: R&D Systems™ RDC0361
Additional Details : Weight : 0.00970kg
Description
The Rat OX40/TNFRSF4 (NP_037181) VersaClone cDNA clone contains the complete ORF for OX40/TNFRSF4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The OX40/TNFRSF4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
OX40/TNFRSF4 | |
10 μg at 0.2 μg/μL | |
828bp | |
ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, O x 40 cell surface antigen, O x 40 homologue, O x 40 L receptor, O x 40 lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LO x 40 antigen | |
Rat |
NP_037181 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
7293 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans