missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-N-BOC-piperidine-3-carboxylic acid, 97%, Thermo Scientific™
1078.00 SEK - 4306.00 SEK
Chemical Identifiers
CAS | 163438-09-3 |
---|---|
Molecular Formula | C11H18NO4 |
Molecular Weight (g/mol) | 228.27 |
MDL Number | MFCD02179173 |
InChI Key | NXILIHONWRXHFA-MRVPVSSYSA-M |
Synonym | r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh |
PubChem CID | 643516 |
SMILES | CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O |
Description
Chemical Identifiers
163438-09-3 | |
228.27 | |
NXILIHONWRXHFA-MRVPVSSYSA-M | |
643516 |
C11H18NO4 | |
MFCD02179173 | |
r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh | |
CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O |
Specifications
163438-09-3 | |
100.0 | |
White | |
96% min. (HPLC) | |
MFCD02179173 | |
-52.2 | |
NXILIHONWRXHFA-MRVPVSSYSA-M | |
(3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid | |
643516 | |
97% | |
(R)-N-BOC-Piperidine-3-carboxylic acid |
96.0 | |
166°C to 171°C | |
Authentic | |
C11H18NO4 | |
−52.20° (20°C c=1.15 MeOH) | |
r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh | |
CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O | |
228.27 | |
229.28 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (R)-N-BOC-Piperidine-3-carboxylic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(R)-N-BOC-piperidine-3-carboxylic acid, 97%, Thermo Scientific™