missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies R848

Immune modulator; Inhibits TLR7 and TLR8

Encompass_Preferred

1583.55 SEK - 5719.95 SEK

Specifications

CAS 144875-48-9
Synonym Resiquimod; S 28463
Molecular Formula C17H22N4O2
Purity ≥98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17117421
Encompass_Preferred
View Documents
STEMCELL Technologies
73782
10 mg
1583.55 SEK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17160272
Encompass_Preferred
View Documents
STEMCELL Technologies
73784
50 mg
5719.95 SEK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • R848 is an imidazoquinoline and agonist of Toll-like receptors (TLRs) 7 and 8
  • It mimics the pathogen-associated molecular patterns that activate immune cells through TLR7 and TLR8, and thereby acts as an immune-response modifier
  • It demonstrates potent anti-tumor and anti-viral properties (IC50 = 4.2 μM; Seganish et al.) which appear to be mediated predominantly through the induction of cytokines, including interferon (IFN)-α and interleukin (IL)-12 due to stimulation of monocytes, macrophages, and dendritic cells (Bernstein et al.; Hattermann et al.; Nian et al.)
Specifications

Specifications

144875-48-9
Resiquimod; S 28463
C17H22N4O2
≥98%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
STEMCELL Technologies R848

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.