Learn More
RAB10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27094-1-AP-150UL
This item is not returnable.
View return policy
Description
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.Specifications
RAB10 | |
Polyclonal | |
Unconjugated | |
RAB10 | |
RAB10, Ras related protein Rab 10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10890 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P61026 | |
RAB10 | |
RAB10 Fusion Protein Ag25830 | |
150 μL | |
Primary | |
Human | |
Antibody | |