missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab11A Mouse anti-Human, Mouse, Rat, Clone: 918009, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8978

This item is not returnable.
View return policy
Description
Rab11A Monoclonal specifically detects Rab11A in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Rab11A | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC1490, RAB 11A, member oncogene family, Rab-11, RAB11, RAB11A, member RAS oncogene family, YL8, YL8ras-related protein Rab-11A | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8766 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot | |
918009 | |
Western Blot 1 ug/mL | |
P62491 | |
RAB11A | |
E. coli-derived recombinant human Rab-11A Met1-Glu162 Accession # P62491 | |
100 μg | |
Primary | |
Detects human Rab-11A in direct ELISAs and Western blots. | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Rab11A Mouse anti-Human, Mouse, Rat, Clone: 918009, R&D Systems™
Spot an opportunity for improvement?Share a Content Correction