missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen A16143
This item is not returnable.
View return policy
Description
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Goat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
FITC | |
Rabbit | |
Affinity chromatography | |
RUO | |
Goat | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |