missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Rack
Brand: Tenak™ TE21430
This item is not returnable.
View return policy
Description
freezer, for 14 boxes, 50mm high, stainless steel, with locking rod, TenakSpecifications
Stainless Steel | |
1 x 14 | |
140 mm | |
140 mm |
Standard sized boxes with 9 x 9 and 10 x 10 cell dividers with a max. footprint of 134 x 134 mm | |
140 x 140 x 775 mm | |
775 mm | |
Cryoboxes (14, 50 mm H) |