missing translation for 'onlineSavingsMsg'
Learn More

RACK1/GNB2L1 Rabbit anti-Human, Mouse, Rat, Clone: S02-1K6, Novus Biologicals™

Rabbit Monoclonal Antibody

2825.00 SEK - 5705.00 SEK

Specifications

Antigen RACK1/GNB2L1
Clone S02-1K6
Dilution Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18331865
View Documents
Bio-Techne
NBP3-14967-25UL
25 μg
2825.00 SEK
25µL
Please to purchase this item. Need a web account? Register with us today!
18343154
View Documents
Bio-Techne
NBP3-14967-100UL
100 μg
5705.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RACK1/GNB2L1 Monoclonal antibody specifically detects RACK1/GNB2L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

RACK1/GNB2L1
Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
guanine nucleotide binding, guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1, Receptor for activated C kinase, Receptor for Activated C Kinase 1
A synthetic peptide of human RACK1/GNB2L1 (Uniprot#: P63244)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
S02-1K6
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Stem Cell Markers
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
10399
IgG
Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.