Learn More
Eppendorf™ RackScan
RackScan reads complete storage racks with up to 96 different barcoded vials. The sample IDs are transferred to downstream databases.
Brand: Eppendorf™ 6002000006
112717.50 SEK valid until 2024-12-20
Use promo code "21731" to get your promotional price.
This item is not returnable.
View return policy
Description
- Camera based instrument resulting in rapid imaging and decoding
- Comfortable and easy plug-in accessory
- Small footprint, ideal for benchtop (and integration)
- ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
- Support material of vials available via Eppendorf Homepage for documentation purposes
- Plug & Transfer with Eppendorf™ eLABInventory and eLABJournal software for comfortable data transfer
- RackScan s can be attached to RackScan b as accessory to read barcodes on side labels
- Different formats usable for high flexibility
Specifications
RackScan b | |
PC/USB | |
Scanning more than one vial in parallel and transferring the sample data to your sample management software | |
RackScan Analyzer Software, USB Cable | |
Scanner (Bottom Reading) | |
LED | |
2D |
Machine-readable | |
2014/35/EU:EN61010-1, 2014/30/EU: EN61326-1, EN 55032, EN61000-3-2, EN 61000-3-3, 2011/65/EU:EN50581 | |
2D-coded tubes in SBS racks and cryostorage boxes | |
RackScan Analyzer Software | |
USB | |
100/240 V |