missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1680.00 SEK - 3415.00 SEK
Specifications
Antigen | Rad21 |
---|---|
Dilution | Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin |
Applications | ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18323514
|
Bio-Techne
NBP3-15608-20UL |
20 μg |
1680.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18336436
|
Bio-Techne
NBP3-15608-100UL |
100 μg |
3415.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rad21 Polyclonal antibody specifically detects Rad21 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
Rad21 | |
ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair | |
PBS, 50% glycerol, pH7.3 | |
5885 | |
IgG | |
Affinity purified |
Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
double-strand-break repair protein rad21 homolog, hHR21FLJ25655, HR21, KIAA0078RAD21 (S. pombe) homolog, MCD1, Nuclear matrix protein 1, NXP-1, NXP1HRAD21, protein involved in DNA double-strand break repair, RAD21 homolog (S. pombe), SCC1 homolog, SCC1FLJ40596 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 268-429 of human Rad21 (NP_006256.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title