Learn More
Rad23 Rabbit anti-Human, Mouse, Rat, Clone: 3B7T9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16573-20UL
This item is not returnable.
View return policy
Description
Rad23 Monoclonal antibody specifically detects Rad23 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Rad23 | |
Monoclonal | |
Unconjugated | |
hHR23A, HR23A | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Rad23 (P54725). | |
20 μg | |
Apoptosis, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
5886 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
3B7T9 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |