missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD52 Monoclonal Antibody (5H9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA123334
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa whole cell lysate.
DNA double-strand breaks can be generated by exposure to ionizing radiation or through endogenous cellular reactions. Double-strand breaks are generally thought to be repaired via one of two distinct mechanisms: homologous recombination (HR) or non-homologous end joining (NHEJ). Homologous recombination requires the activity of the members of the RAD52 epistasis group, which includeRAD50, RAD51, RAD52, RAD54, RDH54/TID1, RAD55, RAD57, RAD59, MRE11, and XRS2. RAD52 can catalyze strand annealing reactions, and can form oligomers that attach to ends of single-stranded DNA.Specifications
RAD52 | |
Monoclonal | |
1 mg/mL | |
PBS with 20% glycerol and no preservative | |
P43351 | |
RAD52 | |
Human Rad52 recombinant protein expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
5H9 | |
Unconjugated | |
RAD52 | |
DNA repair protein RAD52 homolog; RAD52; RAD52 homolog (S. cerevisiae); RAD52 homolog, DNA repair protein; Rad52yh; recombination protein RAD52; rhabdomyosarcoma antigen MU-RMS-40.23 | |
Mouse | |
Protein G | |
RUO | |
5893 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction