Learn More
OHAUS™ Ranger™ 7000
OHAUS™ Ranger™ 7000 boasts a high resolution, has a large display, a variety of connectivity options, and a large memory.
16390.00 SEK - 41715.00 SEK
Specifications
Connection Type | Easy access communication port including standard USB and RS232 (included) and an optional third port of RS232 or Ethernet (accessories sold separately). GLP/GMP Data Output with Real Time Clock. Data Transfer Function. |
---|---|
Construction | Modular design, die-cast housing, removable stainless steel platform, integral weigh below hook, menu lock switch, level indicator |
Display Type | Full-color WQVGA Graphic Display with user-controlled brightness |
Protection Class | IP54 |
Weighing Modes | Gram, Kilogram, Ounce, Pound, Pound:Ounce, Custom |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15614553
|
OHAUS™
30088847 |
35.00 kg |
16390.00 SEK
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16002415
|
OHAUS™
30088832 |
3.00 kg |
41715.00 SEK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Simplify complex applications and minimize the need for manual calculations with 10 advanced application modes, peripheral device control and an optional kit for a second scale platform.
- The most precise scale in its class with one-second stabilization time, up to 75,000d or 350,000d display resolution and legal-for-trade certifications.
- Extremely durable scale designed to thrive in rugged industrial environments.
- Features IP54-rated metal housing and sealed metal terminal.
Specifications
Easy access communication port including standard USB and RS232 (included) and an optional third port of RS232 or Ethernet (accessories sold separately). GLP/GMP Data Output with Real Time Clock. Data Transfer Function. | |
Full-color WQVGA Graphic Display with user-controlled brightness | |
Gram, Kilogram, Ounce, Pound, Pound:Ounce, Custom |
Modular design, die-cast housing, removable stainless steel platform, integral weigh below hook, menu lock switch, level indicator | |
IP54 |