missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ras Rabbit anti-Human, Mouse, Rat, Clone: 3G8L8, Novus Biologicals™
Rabbit Monoclonal Antibody
1950.00 SEK - 4805.00 SEK
Specifications
Antigen | Ras |
---|---|
Clone | 3G8L8 |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18322552
|
Bio-Techne
NBP3-15909-20UL |
20 μg |
1950.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18301832
|
Bio-Techne
NBP3-15909-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ras Monoclonal antibody specifically detects Ras in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
Ras | |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Signaling, Epigenetics | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
IgG | |
Affinity purified |
3G8L8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
A synthesized peptide derived from human KRAS+HRAS+NRAS | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title