Learn More
R&D Systems™ Rat Somatostatin R2/SSTR2 (NP_062221) VersaClone cDNA
Somatostatin Receptor Type 2
Brand:  R&D Systems™ RDC1932
Product Code. 18793559
  • 3340.00 SEK / 10µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
The Rat Somatostatin R2/SSTR2 (NP_062221) VersaClone cDNA clone contains the complete ORF for Somatostatin R2/SSTR2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Somatostatin R2/SSTR2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
cDNA
NP_062221
cDNA is provided in 10mM Tris-Cl, pH 8.5
6752 (Human); 20606 (Mouse); 54305 (Rat)
Rat
rSstr2
10 μg at 0.2 μg/μL
1123bp
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.