missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA556543
This item is not returnable.
View return policy
Description
Immunogen sequence: VTWDEYNIQM YDRVIDFDEN TALDDAEEES FRKLHLKDKK RFEKANQDSG PGLSLEEFIA FEHPEEVDYM TEF Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
The protein encoded by this gene is a calcium-binding protein located in the lumen of the ER. The protein contains six conserved regions with similarity to a high affinity Ca(+2)-binding motif, the EF-hand. This gene maps to the same region as type 4 Bardet-Biedl syndrome, suggesting a possible causative role for this gene in the disorder. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
RCN2 | |
Polyclonal | |
Unconjugated | |
RCN2 | |
AA408742; Calcium-binding protein ERC-55; E6-binding protein; E6BP; ERC55; ERC-55; RCN2; reticulocalbin 2; reticulocalbin 2, EF-hand calcium binding domain; reticulocalbin 2, EF-hand calcium binding domain (endoplasmic reticulum calcium-binding protein, 55kD); reticulocalbin-2; taipoxin-associated calcium binding protein 49; taipoxin-associated calcium-binding protein 49; TCBP49; TCBP-49 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26611, 5955 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14257, Q8BP92 | |
RCN2 | |
Recombinant protein corresponding to Human RCN2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction