Learn More
Gibco™ Recharge™ Supplement, Without Glucose and L-glutamine
A chemically defined, animal origin-free (AOF) protein-free supplement formulated without glucose and L-glutamine.
Brand:  Gibco™ 670002
 View more versions of this product
Product Code. 16209901
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • A chemically defined (CD) supplement designed to increase protein yields, while maintaining acceptable viability and cell growth
  • The supplement has been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors
  • In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations
  • Therefore, these two components should be supplemented by the user and monitored as appropriate
  • The Recharge™ Supplement was developed for use in CHO mammalian cell culture systems and can be used either as a media supplement or a culture system feed
  • However, user data have shown this CD supplement has been beneficial for non-CHO mammalian systems and microbial systems
  • Additionally, the Recharge™ Supplement has been successfully used as a replacement for yeast extract
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store in cool, dry conditions in well-sealed containers, 2°C to 8°C
CHO Media Supplement
100 g
Powder
Mammalian Cell Culture
Animal origin-free