missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Endolysin Protein
A DNA sequence encoding the Enterobacteria phage T4 (Bacteriophage T4) Endolysin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7339-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_049736.1 | |
Endolysin Protein | |
Research Use Only | |
E | |
Recombinant Protein | |
E. coli | |
MNIFEMLRID EGLRLKIYKD TEGYYTIGIG HLLTKSPSLN AAKSELDKAI GRNCNGVITK DEAEKLFNQD VDAAVRGILR NAKLKPVYDS LDAVRRCALI NMVFQMGETG VAGFTNSLRM LQQKRWDEAA VNLAKSRWYN QTPNRAKRVI TTFRTGTWDA YKNL | |
Greater than 90% as determined by SDS-PAGE. |
1258585 | |
100 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction