missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Cytomegalovirus Viral interleukin-10 homolog Protein

A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) Viral interleukin-10 homolog, was expressed in the hosts and tags indicated.

5195.00 SEK - 44210.00 SEK

Specifications

Name Viral interleukin-10 homolog Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol UL111A
Protein Length Full Length
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16087310
View Documents
enQuireBio™
QP7080-10ug
10 μg
5195.00 SEK
10µg
Please to purchase this item. Need a web account? Register with us today!
16027320
View Documents
enQuireBio™
QP7080-50ug
50 μg
7415.00 SEK
50µg
Please to purchase this item. Need a web account? Register with us today!
16077310
View Documents
enQuireBio™
QP7080-100ug
100 μg
12630.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16007320
View Documents
enQuireBio™
QP7080-200ug
200 μg
20825.00 SEK
200µg
Please to purchase this item. Need a web account? Register with us today!
16017320
View Documents
enQuireBio™
QP7080-500ug
500 μg
27820.00 SEK
500µg
Please to purchase this item. Need a web account? Register with us today!
16097310
View Documents
enQuireBio™
QP7080-1mg
1 mg
44210.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Viral interleukin-10 homolog Protein
Not determined.
Full Length
Cytomegalovirus
His-SUMO
Tris-based buffer, 50% glycerol
Research Use Only
UL111A
Recombinant Protein
E. coli
SEEAKPATTT IKNTKPQCRP EDYATRLQDL RVTFHRVKPT LQREDDYSVW LDGTVVKGCW GCSVMDWLLR RYLEIVFPAG DHVYPGLKTE LHSMRSTLES IYKDMRQCPL LGCGDKSVIS RLSQEAERKS DNGTRKGLSE LDTLFSRLEE YLHSRK
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.