missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog BCHE / Butyrylcholinesterase Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) BCHE / Butyrylcholinesterase, was expressed in the hosts and tags indicated.
3180.00 SEK - 40175.00 SEK
Specifications
Name | BCHE / Butyrylcholinesterase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | BCHE |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15937169
|
enQuireBio™
QP5708-ec-10ug |
10 μg | N/A | ||||||
15977169
|
enQuireBio™
QP5708-ec-50ug |
50 μg | N/A | ||||||
15927169
|
enQuireBio™
QP5708-ec-100ug |
100 μg |
11590.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15957169
|
enQuireBio™
QP5708-ec-200ug |
200 μg | N/A | ||||||
15967169
|
enQuireBio™
QP5708-ec-500ug |
500 μg | N/A | ||||||
15947169
|
enQuireBio™
QP5708-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
BCHE / Butyrylcholinesterase Protein | |
Not determined. | |
Full Length | |
Dog | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
BCHE | |
Recombinant Protein | |
NTDQSFPGFP GSEMWNPNTD LSEDCLYLNV WIPTPKPKNA TVMIWIYGGG FQTGTSSLPV YDGKFLARVE RVIVVSVNYR VGALGFLALP GNPEAPGNLG LFDQQLALQW VQKNIAAFGG NPKSVTLFGE SAGAGSVGLH L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title