missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog Caveolin-1 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Caveolin-1, was expressed in the hosts and tags indicated.

3490.00 SEK - 31480.00 SEK

Specifications

Accession Number NP_001003296.1
Gene ID (Entrez) 403980
Name Caveolin-1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16017491
View Documents Promotion Details
enQuireBio™
QP9031-YE-100UG
100 μg
12410.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16057491
View Documents Promotion Details
enQuireBio™
QP9031-YE-500UG
500 μg
29995.00 SEK
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16027491
View Documents Promotion Details
enQuireBio™
QP9031-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16047491
View Documents Promotion Details
enQuireBio™
QP9031-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16067491
View Documents Promotion Details
enQuireBio™
QP9031-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16037491
View Documents Promotion Details
enQuireBio™
QP9031-YE-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001003296.1
Caveolin-1 Protein
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
403980
Research Use Only
CAV1
Recombinant Protein
Yeast
MSGGKYVDSE GHLYTVPIRE QGNIYKPNNK AMAEEMSEKQ VYDAHTKEID LVNRDPKHLN DDVVKIDFED VIAEPEGTHS FDGIWKASFT TFTVTKYWFY RLLSALFGIP MALIWGIYFA ILSFLHIWAV VPCIKSFLIE IQCISRVYSI YVHTFCDPFF EAVGKIFSNI RINMQKET
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications