missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 30S ribosomal protein S7 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8611-ec-10ug

 View more versions of this product

Product Code. 16025561

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_417800.1
30S ribosomal protein S7 Protein
Research Use Only
rpsG
Recombinant Protein
E. coli
PRRRVIGQRK ILPDPKFGSE LLAKFVNILM VDGKKSTAES IVYSALETLA QRSGKSELEA FEVALENVRP TVEVKSRRVG GSTYQVPVEV RPVRRNALAM RWIVEAARKR GDKSMALRLA NELSDAAENK GTAVKKREDV HRMAEANKAF AHYRWLSLRS FSHQAGASSK QPALGYLN
Greater than 90% as determined by SDS-PAGE.
947846
10 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers