missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Beta-lactamase TEM Protein

A DNA sequence encoding the Escherichia coli (strain K12) Beta-lactamase TEM, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7256-ye-100ug

 View more versions of this product

Product Code. 16017890

  • 12935.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_943295.1
Beta-lactamase TEM Protein
Research Use Only
bla
Recombinant Protein
Yeast
HPETLVKVKD AEDQLGARVG YIELDLNSGK ILESFRPEER FPMMSTFKVL LCGAVLSRVD AGQEQLGRRI HYSQNDLVEY SPVTEKHLTD GMTVRELCSA AITMSDNTAA NLLLTTIGGP KELTAFLHNM GDHVTRLDRW EPELNEAIPN DERDTTMPAA MATTLRKLLT GELLTLASRQ QLIDWMEADK VAGPLLRSAL PAGWFIADKS GAGERGSRGI IAALGPDGKP SRIVVIYTTG SQATMDERNR QIAEIGASLI KHW
Greater than 90% as determined by SDS-PAGE.
10076131
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.