missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Beta-lactamase TEM Protein
A DNA sequence encoding the Escherichia coli (strain K12) Beta-lactamase TEM, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7256-ye-200ug
This item is not returnable.
View return policy
Specifications
NP_943295.1 | |
Beta-lactamase TEM Protein | |
Research Use Only | |
bla | |
Recombinant Protein | |
Yeast | |
HPETLVKVKD AEDQLGARVG YIELDLNSGK ILESFRPEER FPMMSTFKVL LCGAVLSRVD AGQEQLGRRI HYSQNDLVEY SPVTEKHLTD GMTVRELCSA AITMSDNTAA NLLLTTIGGP KELTAFLHNM GDHVTRLDRW EPELNEAIPN DERDTTMPAA MATTLRKLLT GELLTLASRQ QLIDWMEADK VAGPLLRSAL PAGWFIADKS GAGERGSRGI IAALGPDGKP SRIVVIYTTG SQATMDERNR QIAEIGASLI KHW | |
Greater than 90% as determined by SDS-PAGE. |
10076131 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction